MAL-dPEG®12-NHS ester

10284-100MG 10284-1000MG
102855-588EA 893.15 USD
102855-588 102855-586
MAL-dPEG®12-NHS ester
MAL-PEG12-NHS ester

Single compound - dPEG® Spacer is 46 atoms and 53.3 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₃₈H₆₃N₃O₁₉
MW: 865.92 g/mol
CAS Number: 756525-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR