Amino-dPEG®8-t-butyl ester

Synonyms: tert-Butyl 1-amino-3,6,9,12,15,18,21,24-octaoxaheptacosan-27-oate

10271-100MG 10271-1000MG
102855-550EA 240.62 USD
102855-550 102855-552
Amino-dPEG®8-t-butyl ester
Amino-PEG8-t-butyl ester

Single compound - dPEG® Spacer is 28 atoms and 32.2 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₂₃H₄₇NO₁₀
MW: 497.62 g/mol
MDL Number: MFCD11041142
CAS Number: 756526-06-4
https://us.vwr.com/stibo/web/std.lang.all/06/41/12380641.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR