CBZ-N-amido-dPEG®3-amine

Synonyms: CBZ-N-amido-dPEG®3-amine

10269-1000MG
102855-548EA 527.54 USD
102855-548
CBZ-N-amido-dPEG®3-amine
CBZ-N-amido-PEG3-amine

Single compound - dPEG® Spacer is 15 atoms and 16.9 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₁₈H₃₀N₂O₅
MW: 354.44 g/mol
CAS Number: 220156-99-0
https://us.vwr.com/stibo/web/std.lang.all/06/39/12380639.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR