Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Biotin-dPEG®3-benzophenone


10267-25MG
102855-540EA 163.46 USD InStock
102855-540
Biotin-dPEG®3-benzophenone
Biotin-dPEG(3)-Benzophenone
The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.


Formula: C₃₄H₄₆N₄O₇S
MDL Number: MFCD13184950
CAS Number: 756525-96-9
EINECS: 000-000-0
https://us.vwr.com/stibo/web/std.lang.all/06/36/12380636.jpg
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty