Amino-dPEG®2-t-butyl ester

Synonyms: 3-[2-(2-Aminoethoxy)ethoxy]propanoic acid tert-butyl estertert-Butyl 3-[2-(2-Aminoethoxy)ethoxy]propanoateAmino-dPEG®2-t-butyl estertert-Butyl 9-Amino-4,7-dioxanonanoate9-Amino-4,7-dioxanonanoic acid tert-butyl ester

10264-100MG 10264-1000MG
102855-530EA 199.92 USD
102855-530 102855-528
Amino-dPEG®2-t-butyl ester
Amino-PEG2-acid tert-butyl ester

Single compound - dPEG® Spacer is 10 atoms and 10.9 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₁₁H₂₃NO₄
MW: 233.305 g/mol
MDL Number: MFCD11041135
CAS Number: 756525-95-8
https://us.vwr.com/stibo/web/std.lang.all/06/30/12380630.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR