m-dPEG®12-NHS ester

Synonyms: Methyl-PEG12-NHS Esterm-dPEG®12-NHS ester

10262-1000MG 10262-100MG
102855-522EA 823.85 USD
102855-522 102855-520
m-dPEG®12-NHS ester
m-PEG12-NHS ester

Single compound - dPEG® Spacer is 38 atoms and 44 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₃₀H₅₅NO₁₆
MW: 685.755 g/mol
MDL Number: MFCD13184970
CAS Number: 756525-94-7
https://us.vwr.com/stibo/web/std.lang.all/06/28/12380628.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR