m-dPEG®11-alcohol

10252-1000MG 10252-100MG
102855-502EA 77.25 USD
102855-502 102855-504
m-dPEG®11-alcohol
m-PEG11-alcohol

Single compound - dPEG® Spacer is 35 atoms and 40.3 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₂₃H₄₈O₁₂
MW: 516.62 g/mol
CAS Number: 114740-40-8
https://us.vwr.com/stibo/web/std.lang.all/05/25/12380525.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR