Amino-dPEG®4-alcohol

10249-100MG 10249-1000MG
102855-492EA 206.62 USD
102855-492 102855-490
Amino-dPEG®4-alcohol
Amino-PEG4-alcohol

Single compound - dPEG® Spacer is 13 atoms and 14.3 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₈H₁₉NO₄
MW: 193.24 g/mol
CAS Number: 86770-74-3
https://us.vwr.com/stibo/web/std.lang.all/05/19/12380519.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR