Bis-dPEG®5, half benzyl half NHS ester

Synonyms: Bis-dPEG®5, half benzyl half NHS ester

10237-100MG 10237-1000MG
102855-462EA 343.92 USD
102855-462 102855-464
Bis-dPEG®5, half benzyl half NHS ester
Bis-PEG5, half benzyl half NHS ester

Single compound - dPEG® Spacer is 19 atoms and 21.7 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₂₅H₃₅NO₁₁
MW: 525.55 g/mol
CAS Number: 1263044-84-3
https://us.vwr.com/stibo/web/std.lang.all/06/20/12380620.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR