m-dPEG®4-acid
10234-1000MG
10234-100MG
102855-456EA
363.54
USD
102855-456
102855-454
m-dPEG®4-acid
m-PEG4-acid
Single compound - dPEG® Spacer is 14 atoms and 15.6 Å
The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.
MW:
236.26 g/mol |
CAS Number:
67319-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...