Hydroxy-dPEG®4-t-butyl ester

10223-100MG 10223-1000MG
102855-434EA 274.62 USD
102855-434 102855-432
Hydroxy-dPEG®4-t-butyl ester
Hydroxy-PEG4-t-butyl ester

Single compound - dPEG® Spacer is 16 atoms and 18.0 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₁₅H₃₀O₇
MW: 332.39 g/mol
CAS Number: 518044-32-1
https://us.vwr.com/stibo/web/std.lang.all/18/80/12381880.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR