Biotin-dPEG®4-hydrazide

Synonyms: Hydrazide-PEG4-Biotin15-(Biotinamido)-4,7,10,13-tetraoxapentadecanehydrazideN-[2-[2-[2-[2-(3-Hydrazinyl-3-oxopropoxy)ethoxy]ethoxy]ethoxy]ethyl]biotinamide

10219-50MG 10219-1000MG
102855-420EA 1167.77 USD
102855-420 102855-418
Biotin-dPEG®4-hydrazide
Biotin-PEG4-hydrazide

Single compound - dPEG® Spacer is 18 atoms and 20.6 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₂₁H₃₉N₅O₇S
MW: 505.63 g/mol
MDL Number: MFCD09952658
CAS Number: 756525-97-0
https://us.vwr.com/stibo/web/std.lang.all/06/04/12380604.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR