Bis-MAL-dPEG®3

Synonyms: Bis-MAL-dPEG®3

10215-50MG 10215-1000MG
102855-410EA 1030.46 USD
102855-410 102855-408
Bis-MAL-dPEG®3
Bis-Mal-PEG3

Single compound - dPEG® Spacer is 28 atoms and 30 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₂₂H₃₀N₄O₉
MW: 494.495 g/mol
MDL Number: MFCD22683311
CAS Number: 756525-89-0
https://us.vwr.com/stibo/web/std.lang.all/05/99/12380599.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR