Fmoc-N-amido-dPEG®4-acid

10213-1000MG 10213-5000MG 10213-100MG
102855-400EA 234.08 USD
102855-400 102855-402 102855-398
Fmoc-N-amido-dPEG®4-acid
Fmoc-N-amido-dPEG®4-acid

Single compound - dPEG® Spacer is 17 atoms and 18.1 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₂₆H₃₃NO₈
MW: 487.54 g/mol
CAS Number: 557756-85-1
https://us.vwr.com/stibo/web/std.lang.all/05/95/12380595.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR