NHS-dPEG®4-biotinidase resistant biotin

Synonyms: NHS-dPEG®4-biotinidase resistant biotin

10202-1000MG 10202-100MG
102855-378EA 206.62 USD
102855-378 102855-380
NHS-dPEG®4-biotinidase resistant biotin
NHS-PEG4-biotinidase resistant biotin

Single compound - dPEG® Spacer is 19 atoms and 21.5 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₂₉H₄₇N₅O₁₁S
MW: 673.78 g/mol
CAS Number: 1334172-61-0
https://us.vwr.com/stibo/web/std.lang.all/05/84/12380584.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR