NHS-dPEG®4-biotin

10200-1000MG 10200-50MG
102855-374EA 1030.46 USD
102855-374 102855-372
NHS-dPEG®4-biotin
NHS-PEG4-biotin

Single compound - dPEG® Spacer is 16 atoms and 19.2 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₂₅H₄₀N₄O₁₀S
MW: 588.67 g/mol
CAS Number: 459426-22-3
https://us.vwr.com/stibo/web/std.lang.all/05/81/12380581.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR