NHS-dPEG®12-biotin

10198-50MG 10198-1000MG
102855-364EA 171.31 USD
102855-364 102855-366
NHS-dPEG®12-biotin
NHS-PEG12-biotin

Single compound - dPEG® Spacer is 40 atoms and 47.6 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₄₁H₇₂N₄O₁₈S
MW: 941.09 g/mol
CAS Number: 365441-71-0
https://us.vwr.com/stibo/web/std.lang.all/05/77/12380577.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR