dPEG®12-biotin acid

Synonyms: dPEG®12-biotin acid

10197-1000MG 10197-100MG
102855-360EA 206.62 USD
102855-360 102855-362
dPEG®12-biotin acid
PEG12-biotin acid

Single compound - dPEG® Spacer is 40 atoms and 47.6 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₃₇H₆₉N₃O₁₆S
MW: 844.02 g/mol
CAS Number: 948595-11-7
https://us.vwr.com/stibo/web/std.lang.all/05/75/12380575.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR