Biotin-dPEG®11-NH2

Synonyms: Biotin-PEG11-Amine

10196-1000MG 10196-100MG
102855-356EA 377.92 USD
102855-356 102855-358
Biotin-dPEG®11-NH2
Biotin-PEG11-NH2

Single compound - dPEG® Spacer is 37 atoms and 44.1 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₃₄H₆₆N₄O₁₃S
MW: 770.97 g/mol
CAS Number: 604786-74-5
https://us.vwr.com/stibo/web/std.lang.all/05/73/12380573.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR