Biotin-dPEG®11-MAL

10195-100MG 10195-25MG
102855-352EA 302.08 USD
102855-352 102855-354
Biotin-dPEG®11-MAL
Biotin-PEG11-MAL

Single compound - dPEG® Spacer is 43 atoms and 50.5 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₄₁H₇₁N₅O₁₆S
MW: 922.09 g/mol
CAS Number: 1334172-60-9
https://us.vwr.com/stibo/web/std.lang.all/05/71/12380571.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR