Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Biotin-dPEG®3-NH3+TFA-

Supplier: QUANTA BIODESIGN MS

10193-100MG 10193-1000MG
102855-344EA 130.77 USD InStock
102855-344 102855-346
Biotin-dPEG®3-NH3+TFA-
Biotin-dPEG®3-NH3+TFA-
Single compound - dPEG® Spacer is 15 atoms and 18.1 Å

The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.


Formula: C₂₂H₃₉F₃N₄O₇S
MW: 560,63 g/mol
CAS Number: 1334172-59-6
https://us.vwr.com/stibo/web/std.lang.all/05/67/12380567.jpg
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty