MPS-EDA.TFA

10177-1000MG 10177-100MG
102855-312EA 303.38 USD
102855-312 102855-314
MPS-EDA.TFA
MPS-EDA.TFA

Single compound - dPEG® Spacer is 10 atoms and 10.7 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₁₁H₁₄F₃N₃O₅
CAS Number: 11550-02-00

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR