m-dPEG®4-amine

10175-100MG 10175-1000MG
102855-310EA 253.69 USD
102855-310 102855-308
m-dPEG®4-amine
m-PEG4-amine

Single compound - dPEG® Spacer is 14 atoms and 15.5 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₉H₂₁NO₄
MW: 207.27 g/mol
CAS Number: 85030-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR