Amino-dPEG®12-alcohol

10170-1000MG 10170-100MG
102855-298EA 893.15 USD
102855-298 102855-296
Amino-dPEG®12-alcohol
Amino-PEG12-alcohol

Single compound - dPEG® Spacer is 38 atoms and 42.7 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₂₄H₅₁NO₁₂
MW: 545.66 g/mol
MDL Number: MFCD11041097
CAS Number: 933789-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR