Acid-dPEG®13-NHS ester

Synonyms: Acid-dPEG®13-NHS ester

10127-100MG 10127-1000MG
102855-282EA 1785 USD
102855-282 102855-280
Acid-dPEG®13-NHS ester
Acid-PEG13-NHS ester

Single compound - dPEG® Spacer is 43 atoms and 50.0 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₃₄H₆₁NO₁₉
MW: 787.843 g/mol
MDL Number: MFCD27952889
CAS Number: 2152679-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR