Acid-dPEG®5-NHS ester

Synonyms: Acid-dPEG®5-NHS ester

10109-100MG 10109-1000MG
102855-276EA 295.54 USD
102855-276 102855-278
Acid-dPEG®5-NHS ester
Acid-PEG5-NHS ester

Single compound - dPEG® Spacer is 19 atoms and 21.6 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₁₈H₂₉NO₁₁
MW: 435.423 g/mol
MDL Number: MFCD27952888
CAS Number: 1343476-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR