Bis-dPEG®25-TFP ester
Synonyms: Bis-dPEG®25-TFP ester
10092-1000MG 10092-100MG
102855-272EA 481.23 USD
102855-272 102855-274
Bis-dPEG®25-TFP ester
Bis-PEG25-TFP ester
Single compound - dPEG® Spacer is 79 atoms and 93.0 Å
The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.