CBZ-N-amido-dPEG®6-acid

10066-100MG 10066-1000MG
102855-254EA 411.92 USD
102855-254 102855-252
CBZ-N-amido-dPEG®6-acid
CBZ-N-amido-PEG6-acid

Single compound - dPEG® Spacer is 22 atoms and 25.1 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₂₃H₃₇NO₁₀
MW: 487.54 g/mol
CAS Number: 1334177-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR