MAL dPEG®6-acid

Synonyms: 1-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3-oxo-7,10,13,16,19,22-hexaoxa-4-azapentacosan-25-oic acid

10065-100MG 10065-1000MG
102855-248EA 171.31 USD
102855-248 102855-250
MAL dPEG®6-acid
Mal-amido-PEG6-acid

Single compound - dPEG® Spacer is 28 atoms and 31.9 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₂₂H₃₆N₂O₁₁
MW: 504.53 g/mol
MDL Number: MFCD21363276
CAS Number: 1334177-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR