Amino-dPEG®6-t-butyl ester

10061-1000MG 10061-100MG
102855-236EA 481.23 USD
102855-236 102855-234
Amino-dPEG®6-t-butyl ester
Amino-PEG6-t-butyl ester

Single compound - dPEG® Spacer is 22 atoms and 25.1 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₁₉H₃₉NO₈
MW: 409.51 g/mol
CAS Number: 1286281-32-0
https://us.vwr.com/stibo/web/std.lang.all/48/18/12744818.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR