Amino-dPEG®4-t-boc-hydrazide

Synonyms: Amino-dPEG®4-t-boc-hydrazideNH2-PEG4-HZ-Boc

10041-1000MG 10041-100MG
102855-220EA 1030.46 USD
102855-220 102855-218
Amino-dPEG®4-t-boc-hydrazide
Amino-PEG4-t-boc-hydrazide

Single compound - dPEG® Spacer is 18 atoms and 21.1 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₁₆H₃₃N₃O₇
MW: 379.449 g/mol
MDL Number: MFCD11041089
CAS Number: 1263047-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR