Fmoc-N-amido-dPEG®3-acid

10033-1000MG 10033-5000MG
102855-214EA 226.23 USD
102855-214 102855-216
Fmoc-N-amido-dPEG®3-acid
Fmoc-N-amido-dPEG®3-acid

Single compound - dPEG® Spacer is 13 atoms and 14.4 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₂₄H₂₉NO₇
MW: 443.49 g/mol
CAS Number: 867062-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR