Bis-dPEG®5-PFP ester

10015-100MG 10015-1000MG
102855-210EA 154.5 USD
102855-210 102855-212
Bis-dPEG®5-PFP ester
Bis-PEG5-PFP ester

Single compound - dPEG® Spacer is 19 atoms and 21.7 Å


The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.

Formula: C₂₆H₂₄F₁₀O₉
MW: 670.447 g/mol
MDL Number: MFCD18916983
CAS Number: 1334177-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR