Biotin-dPEG®4-TFP ester
10009-1000MG 10009-100MG
102855-198EA 823.85 USD
102855-198 102855-196
Biotin-dPEG®4-TFP ester
Biotin-PEG4-TFP ester
Single compound - dPEG® Spacer is 16 atoms and 19.2 Å
The dPEG® product line is a unique technology platform, single MW PEGs, which can be tailored to meet specific physical and chemical requirements in a broad array of diagnostic, therapeutic and nanotech applications. These applications include conjugations, chemical modifications, cross linking and modification of biological therapeutics. The dPEG® products are available in a range of sizes to meet your specific requirements.