LIBERO Ti1 and Ti1-L USB Ambient and Refrigerated Temperature Monitoring Data Logger, ELPRO
Supplier: ELPRO SERVICES, INC.
800011 800004
75784-778EA 421.83 USD InStock
75784-778 10032-782
LIBERO Ti1 and Ti1-L USB Ambient and Refrigerated Temperature Monitoring Data Logger, ELPRO
Data Loggers
The LIBERO Ti1 and Ti1-L USB Data Loggers are a safe, easy, and inexpensive way to monitor and record a wide range of temperature conditions.
This compliant and economic solution for continuous monitoring is ideal for many applications in areas such as lab freezers, refrigerators, incubators, ambient room conditions, and/or deli cases
All settings, including alarm ranges and logging interval, are user-configurable via a free configuration tool. Choose from high/low or multi-alarm levels with user-defined alarm delay. Once configured, insert it into any computer USB port to generate a 21 CFR Part 11 Compliant PDF report with no software or download cables required. Additionally, end users have access to on-screen statistics such as min./max./average, current temperature, and alarm status. If necessary, point-by-point data can be accessed via a free downloadable tool that allows additional notation, reporting, and optional export of raw data to Excel.
The device features a digital display with a visual alarm status that is easy to see (with an "X" or "check"), temperature status, and remaining battery life. Battery life for the Ti1 is 400 days (1 year) and the Ti1-L is 2 years. At the end of battery life, simply re-order the data logger. The Ti1-L can be recalibrated annually by ELPRO.
The LIBERO complies with the ISO standard 19005-1 document management for the longterm preservation of electronic documents and is 21 CFR Part 11 and WHO compliant. The Ti1/Ti1-L is delivered with a validation certificate. There are no more hassles of recalibration/recertification for the Ti1.
Application temperature: Frozen (–20 °C), Cool (2 to 8 °C), refrigerated/ambient (2 to 25 °C), warm (37 to 70 °C)
Certifications: CE marked. The LIBERO meets FDA 21 CFR Part 11 and is WHO compliant.