LIBERO Te1-P and Te1-PY USB Data Loggers for Ultra-Low Freezer Monitoring, ELPRO
802128 800019
75784-782EA 642.14 USD
75784-782 10032-538
LIBERO Te1-P and Te1-PY USB Data Loggers for Ultra-Low Freezer Monitoring, ELPRO
The LIBERO USB Data Loggers monitor equipment 24/7, and automatically generates a PDF report without the need for software or hardware.
To download the data, simply plug the LIBERO into any USB port on any computer
These LIBERO kits are designed to monitor extremely low temepratures (down to -200C), such as ultra-low freezers or liquid nitrogen tanks. Choose between visual or audible/visual alarm indication. They are delivered ready to install with a NIST-traceable, one-point calibration certificate. Additional calibration points and ISO 17025 calibration certificates are available. Choose between a data logger with a battery life of either 400 days (Te1-PY) or 2 years (Te1-P), and at the end of the battery life, simply order a replacement data logger, which is delivered with a validation certificate.
Implementing the LIBERO USB data logger is easy. Just place the sensor inside the equipment and mount the data logger on the outside of the equipment via velcro or double-sided tape. All settings, including alarm ranges and logging interval, are user-configurable via a free configuration tool. Choose from high/low or mulit-alarm levels with user-defined alarm delay. If necessary, point-by-point data can be accessed via a downloadable tool that allows additional notation, reporting, and optional export of raw data to Excel.
The report contains statistics such as min., max., average, and time/duration of alarm as well as alarm settings and a temperature graph. The alarm status is easy to see, with an X or "check" at the top of the page as well as on the data logger display screen. The LIBERO meets FDA 21 CFR Part 11 and is WHO compliant.
Certifications: CE marked. The LIBERO meets FDA 21 CFR Part 11 and is WHO compliant.
Ordering information: Configuration of kits give you the option of 3 meter sensor, no bracket or alarm bracket, 1 year or 2 year battery life. All kits include NIST calibration certificate at -80 °C.