Would you like to log in again?
Avantor is taking action. Our response to the Novel Coronavirus outbreak.
NewClassic Level, TLE Series Precision Balances, METTLER TOLEDO®
NewClassic
30046476 30046473 30046473 30046476
10031-616EA 2178.11 USD InStock
CA10031-618 CA10031-616 10031-616 10031-618
NewClassic Level, TLE Series Precision Balances, METTLER TOLEDO®
Balances and Scales
TLE Series Precision balances offer effortless cleaning, comfortable daily use, and environmental friendliness. They are the perfect solution for classroom applications.
  • Capacities from 320 to 3200 g
  • Readabilities from 0.2 mg
  • Meets all GLP and ISO 9001 Requirements
NewClassic TLE Balances support teaching and learning
These robust balances are designed to withstand intensive use and to provide years of reliable results.
Teaching Lab products are specifically designed for the needs of the classroom – to make teaching and learning as easy as possible for both teachers and students. Daily weighing tasks are easily executed thanks to the five clear and intuitive function keys. Applications can be accessed at the touch of a button and the large numbers on the brilliant backlit display are easy to read.
Certifications: METTLER TOLEDO® provides ISO 9001 manufacturing, nationwide ISO 9001 field service, ISO 9001 quality certification, and ISO 14001 environmental certification. UL listed. CSA certified.
Ordering information: TLE Series Precision balances are packaged with appropriate power supply.
Order Now
ORDER
SPECIFICATIONS
SERVICES
Services: METTLER TOLEDO® provides several service options, including Installation and Qualification, as well as extended warrantys. To learn more about these services please search "METTLER TOLEDO® Installation and Qualification Services" in the VWR.com search field.
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty