Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
LIBERO Te1-N/NY USB Data Loggers for Refrigerator/Lab Freezer Monitoring, ELPRO
Supplier: ELPRO SERVICES, INC.
810022 810026 810021 810025 800016 810023 810024
10032-526EA 465.86 USD InStock
10032-526 75784-788 10032-528 75784-786 75784-780 10032-534 10032-536
LIBERO Te1-N/NY USB Data Loggers for Refrigerator/Lab Freezer Monitoring, ELPRO
Data Loggers
The LIBERO Te1-N USB Data Logger is a safe, easy, highly accurate, and inexpensive way to monitor and record a wide range of temperature conditions. Applications include precise 24/7 monitoring of equipment, pharmacies and more. The data logger automatically generates a PDF report without the need for software or hardware. The LIBERO complies with the ISO standard 19005-1 document management for the long-term preservation of electronic documents. It is 21 CFR Part 11 and WHO compliant.
The LIBERO USB Data Logger monitors equipment 24/7, and automatically generates a PDF report without the need for software or hardware
These LIBERO kits are designed to monitor lab freezers and refrigerators. Choose between visual or audible/visual alarm indication. The equipment monitoring kits are delivered ready to install with a NIST-traceable, one-point calibration certificate. Additional calibration points and ISO 17025 calibration certificates are available. Choose between a battery life of 400 days or 2 years, and at the end of the battery life, simply order a replacement data logger, which is delivered with a validation certificate.
Implementing the LIBERO USB data logger is easy. Simply place the sensor inside the equipment and mount the data logger on the outside of the equipment via velcro or double-sided tape. All settings, including alarm ranges and logging interval, are user-configurable via a free configuration tool. Choose from high/low or multi-alarm levels with user-defined alarm delay. Additionally, end users have access to on-screen statistics such as min./max./average, current temperature, and alarm status. If necessary, point-by-point data can be accessed via a downloadable tool that allows additional notation, reporting, and optional export of raw data to Excel.
The LIBERO Te1-N/NY Data Logger features the built-in capability to generate PDF reports with embedded raw data. No software is required.
Certifications: CE marked. The LIBERO meets FDA 21 CFR Part 11 and is WHO compliant.
Ordering information: Configuration of kits give you the option of: 3 meter sensor, no bracket or alarm bracket, 1 year or 2 year battery life. All kits include NIST calibration certificate at 0 degrees C.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty