Maltodextrin FCC

M1083-2.5KG M1083-12KG M1083-500GM M1083-45KGBL
700011-324EA 675.83 USD
700011-324 95034-284 95034-282 89078-870
Maltodextrin FCC
Maltodextrin

Maltodextrin, FCC is used as a thickener, filler, or preservative in many processed foods. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₂H₂₂O₁₁
MW: 342.29648 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00146679
CAS Number: 9050-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR