DL-α-Tocopherol, Millipore®

Supplier: MilliporeSigma

Synonyms: Vitamin Eall-rac-α-TocopherolVitamin E Alcohol(±)-α-Tocopherol3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-ol

613420-50GM 613420-50GM
80109-028EA 114.48 USD
80109-028 CA80109-028
DL-α-Tocopherol, Millipore®
DL-α-Tocopherol

A physiological membrane-bound antioxidant that protects cell membrane lipids from oxidative damage by free radicals.

Formula: C₂₉H₅₀O₂
MW: 430.71 g/mol
MDL Number: MFCD00072051
CAS Number: 10191-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR