Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenol : chloroform : iso-amyl alcohol (25:24:1) saturated with 100 mM Tris-EDTA to pH 8.0 stabilized Molecular biology grade DNAse, RNAse and Protease free pH 7.8-8.2

Supplier: ACROS ORGANICS MS

Danger

AC32711-5000 AC32711-0025 AC32711-1000
200058-242EA 303.48 USD InStock
200058-242 103527-644 103527-646
Phenol : chloroform : iso-amyl alcohol (25:24:1) saturated with 100 mM Tris-EDTA to pH 8.0 stabilized Molecular biology grade DNAse, RNAse and Protease free pH 7.8-8.2
Phenol : Chloroform : iso-Amyl alcohol


Density: 1.28
Storage Temperature: Refrigerator
MDL Number: MFCD00133763
CAS Number: 136112-00-0
UN: 2810
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear yellow to orange Solution
Stabilizer 0.08 to 0.12 % hydroxyquinoline
pH 7.8 to 8.2 (at 25°C)
Composition mixture of Phenol/Chloroform/Isoamyl alcohol in the ratio of 25:24:1 (v/v/v)
Dnase,Rnase,Protease activity None detected
Iron (Fe) ≤1 ppm
Heavy metals (as Pb) ≤1 ppm


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty