Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Ethanol absolute ≥99.5% ACS (200 Proof)

Supplier: ACROS ORGANICS MS

Danger
Synonyms: Ethyl alcohol

AC61509-0020 AC61509-0040 AC61509-5000
200004-482EA 209.83 USD InStock
200004-482 200004-484 200004-486
Ethanol absolute ≥99.5% ACS (200 Proof)
Ethanol


Formula: H₃CCH₂OH
MW: 46.07 g/mol
Boiling Pt: 78.3 °C (1013 hPa)
Melting Pt: -117 °C
Density: 0.7895 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II
REACH: 01-2119457610-43
Merck Index: 12,03806
Order Now
Specification Test Results
Boiling Point 78°
Melting Point -114°
Flash Point 12°
Density 0.79 g/cm3


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty