Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroacetamide for Pharma synthesis, Non-GMP

Supplier: MilliporeSigma

Danger
Synonyms: 2-Chloroacetamide

8.45148.0100 8.45148.0100 8.45148.2500 8.45148.2500 8.45148.1000 8.45148.1000
EM8.45148.0100CS 1560.11 USD InStock
EM8.45148.0100 CA8.45148.0100 CA8.45148.2500 EM8.45148.2500 CA8.45148.1000 EM8.45148.1000
Chloroacetamide for Pharma synthesis, Non-GMP
Chloroacetamide


Formula: C₂H₄ClNO
MW: 93.51 g/mol
Boiling Pt: 256 °C (1013 hPa)
Melting Pt: 115 - 117 °C
Density: 1.6 g/cm³ (20 °C)
Flash Pt: 170 °C
Storage Temperature: Ambient
CAS Number: 79-07-2
EINECS: 201-174-2
UN: 2811
ADR: 6.1,III
Merck Index: 13,02127
Order Now
Specification Test Results
Solubility 100 g/l (20 °C)
Melting Point 115 - 118 °C
Molar Mass 93.51 g/mol
Bulk Density 760 kg/m3
Boiling Point 220 °C
Vapor Pressure 0.0026 hPa (20 °C)
Flash Point 170 °C
Density (20 °C) no data available
pH 3.8 - 4.1 (40 g/l, H2O, 20 °C)
Saturation concentration (20 °C) Air,no data available
Hazard Class 6.1
Hazard Class Description Toxic materials


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty