Sodium butyrate for pharma synthesis, non-GMP, Sigma-Aldrich®

Warning

Synonyms: Butyric acid sodium saltSodium Butyrate

M-Clarity™ quality level = MQ400

8.45144.0100 8.45144.0100
EM8.45144.0100CS 590.88 USD
CA8.45144.0100 EM8.45144.0100
Sodium butyrate for pharma synthesis, non-GMP, Sigma-Aldrich®
Sodium butyrate
Formula: C₄H₇NaO₂
MW: 110.09 g/mol
Melting Pt: 244 °C
Density: 1.324 g/cm³ (30 °C)
MDL Number: MFCD00002816
CAS Number: 156-54-7
EINECS: 205-857-6

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 244 °C
Molar Mass 110.09 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR