Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Veratric acid for Pharma synthesis, Non-GMP

Supplier: MilliporeSigma

Synonyms: 3,4-Dimethoxybenzoic acid

8.45140.1000 8.45140.0250 8.45140.0250 8.45140.1000
EM8.45140.1000CS 3147.94 USD InStock
EM8.45140.1000 CA8.45140.0250 EM8.45140.0250 CA8.45140.1000
Veratric acid for Pharma synthesis, Non-GMP
Veratric acid


Formula: C₉H₁₀O₄
MW: 182,18 g/mol
Boiling Pt: 303 °C (1013 hPa)
Melting Pt: 117 - 119 °C
Density: 1,436 g/cm³ (-100 °C)
Flash Pt: ≥ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002500
CAS Number: 93-07-2
EINECS: 202-215-7
Merck Index: 12,10086
Order Now
Specification Test Results
Solubility 0.5 g/l (14 °C)
Melting Point 179 - 182 °C
Molar Mass 182.17 g/mol


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty