Diethylamine for pharma synthesis Non-Gmp, Sigma-Aldrich®

Danger


M-Clarity™ quality level = MQ400

8.45028.0100 8.45028.0100
EM8.45028.0100CS 591.39 USD
EM8.45028.0100 CA8.45028.0100
Diethylamine for pharma synthesis Non-Gmp, Sigma-Aldrich®
Diethylamine
Formula: (C₂H₅)₂NH
MW: 73.14 g/mol
Boiling Pt: 55.5 °C (1013 hPa)
Melting Pt: –50 °C
Density: 0.7048 g/cm³ (20 °C)
Flash Pt: –39 °C
Storage Temperature: Ambient
MDL Number: MFCD00009032
CAS Number: 109-89-7
EINECS: 203-716-3
UN: 1154
ADR: 3 (8),II
Merck Index: 13,03138
https://us.vwr.com/stibo/web/std.lang.all/76/20/2297620.jpg

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -50 °C
Molar Mass 73.14 g/mol
Boiling Point 56 °C
Vapor Pressure 250 hPa (20 °C)
Flash Point -25 °C
Refractive Index 1.3864 (20 °C)
Explosion Limit 2.0 - 11.8 %(V)
Density 0.71 g/cm3 (20 °C)
pH 13 (100 g/l, H2O, 20 °C)
Ignition Point 310 °C
Hazard Class 3
Hazard Class Description Flammable liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR