m-Xylene reference substance for gas chromatography, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: 1,3-Dimethylbenzene

1.09797.0005 1.09797.0005
EM1.09797.0005CS 82683 USD
EM1.09797.0005 CA1.09797.0005
m-Xylene reference substance for gas chromatography, Supelco®
m-Xylene
Formula: C₈H₁₀
MW: 106.17 g/mol
Boiling Pt: 138 °C (1013 hPa)
Melting Pt: –48 °C
Density: 0.864 g/cm³ (20 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00008536
CAS Number: 108-38-3
EINECS: 203-576-3
UN: 1307
ADR: 3,III
Merck Index: 12,10214

Order Now

Specification Test Results

Solubility 0.2 g/l (20 °C)
Melting Point -48 °C
Molar Mass 106.16 g/mol
Boiling Point 139 °C (1013 hPa)
Vapor Pressure 8 hPa (20 °C)
Flash Point 25 °C
Explosion Limit 1.1 - 7 %(V)
Density 0.86 g/cm3 (20 °C)
Ignition Point 525 °C
Saturation concentration 35 g/m3 (20 °C) Air
Evaporation number 13.5
Hazard Class 3
Hazard Class Description Flammable liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR