n-Octane reference substance for gas chromatography, Supelco®

Supplier: MilliporeSigma
Danger

1.09716.0005 1.09716.0005
EM1.09716.0005CS 83538 USD
EM1.09716.0005 CA1.09716.0005
n-Octane reference substance for gas chromatography, Supelco®
n-Octane
Formula: H₃C(CH₂)₆CH₃
MW: 114.23 g/mol
Boiling Pt: 126 °C (1013 hPa)
Melting Pt: –57 °C
Density: 0.703 g/cm³ (20 °C)
Flash Pt: 8 °C
Storage Temperature: Ambient
MDL Number: MFCD00009556
CAS Number: 111-65-9
EINECS: 203-892-1
UN: 1262
ADR: 3,
Merck Index: 12,06847
https://us.vwr.com/stibo/web/std.lang.all/79/71/737971.jpg

Order Now

Specification Test Results

Solubility 0.0007 g/l (20 °C)
Melting Point -57 °C
Molar Mass 114.23 g/mol
Boiling Point 125 - 126 °C (1013 hPa)
Vapor Pressure 14 hPa (20 °C)
Refractive Index 13 °C
Explosion Limit 0.8 - 6.5 %(V)
Density 0.70 g/cm3 (20 °C)
Ignition Point 210 °C
Saturation concentration 70 g/m3 (20 °C) Air
Hazard Class 3
Hazard Class Description Flammable liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR