n-Dodecane, Msynth®plus for synthesis

Danger

1.09658.0005 1.09658.0005
EM1.09658.0005CS 54333.75 USD InStock
EM1.09658.0005 CA1.09658.0005
n-Dodecane, Msynth®plus for synthesis
n-Dodecane
Formula: H₃C(CH₂)₁₀CH₃
MW: 170.34 g/mol
Boiling Pt: 216 °C (1013 hPa)
Melting Pt: -9,6 °C
Density: 0,7487 g/cm³ (20 °C)
Flash Pt: 85,5 °C
Storage Temperature: Ambient
MDL Number: MFCD00008969
CAS Number: 112-40-3
EINECS: 203-967-9
https://us.vwr.com/stibo/web/std.lang.all/61/61/736161.jpg

Order Now

Specification Test Results

Solubility (25 °C) practically insoluble
Melting Point -9.6 °C
Molar Mass 170.34 g/mol
Boiling Point 216.3 °C (1013 hPa)
Vapor Pressure 0.4 hPa (20 °C)
Flash Point 74 °C
Explosion Limit 0.6 - 40 %(V)
Density 0.75 g/cm3 (20 °C)
Ignition Point 200 °C
Viscosity kinematic  - 7 mm2/s (40 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR