Ethylbenzene reference substance for gas chromatography, Supelco®

Danger

Synonyms: Phenylethane

1.09635.0005 1.09635.0005
EM1.09635.0005CS 80625.1 USD
CA1.09635.0005 EM1.09635.0005
Ethylbenzene reference substance for gas chromatography, Supelco®
Ethylbenzene
Formula: C₈H₁₀
MW: 106.17 g/mol
Boiling Pt: 136.2 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00011647
CAS Number: 100-41-4
EINECS: 202-849-4
UN: 1175
ADR: 3,II
Merck Index: 13,03801
https://us.vwr.com/stibo/web/std.lang.all/60/41/766041.jpg

Order Now

Specification Test Results

Solubility 0.2 g/l (20 °C)
Melting Point -95 °C
Molar Mass 106.16 g/mol
Boiling Point 136 °C (1013 hPa)
Vapor Pressure 9.5 hPa (20 °C)
Flash Point 15 °C
Explosion Limit 1.0 - 7.8 %(V)
Density 0.87 g/cm3 (20 °C)
Ignition Point 430 °C
Saturation concentration 40 g/m3 (20 °C) Air
Hazard Class 3
Hazard Class Description Flammable liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR